Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199525.1 | NP_001186454.1 | 2 - 3 | 368 | 146 | |
GenBank mRNA | CR354049.1 | - | 2 - 3 | 274 | 146 | |
CR354102.1 | - | 2 - 3 | 361 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |