Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277342.1 | NP_001264271.1 | 3 - 4 | 317 | 115 | |
NM_001277343.1 | NP_001264272.1 | 3 - 4 | 317 | 115 | ||
NM_001277344.1 | NP_001264273.1 | 3 - 4 | 317 | 115 | ||
XM_015288213.2 | XP_015143699.1 | 3 - 4 | 321 | 115 | ||
GenBank mRNA | BU219490.1 | - | 2 - 3 | 309 | 115 | |
BU220011.1 | - | 3 - 4 | 269 | 116 | ||
BX934192.1 | - | 2 - 3 | 291 | 115 | ||
BX934456.2 | - | 3 - 4 | 303 | 115 | ||
BX936012.1 | - | 3 - 4 | 299 | 115 | ||
CR386795.1 | - | 2 - 3 | 289 | 115 | ||
DR430568.1 | - | 3 - 4 | 300 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |