Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252276.2 | NP_001239205.1 | 2 - 3 | 1829 | 104 | |
XM_015285143.2 | XP_015140629.1 | 3 - 4 | 3247 | 104 | ||
XM_015285150.2 | XP_015140636.1 | 3 - 4 | 3061 | 104 | ||
XM_025148538.1 | XP_025004306.1 | 4 - 5 | 3126 | 104 | ||
GenBank mRNA | CR387914.1 | - | 1 - 2 | 540 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |