Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151997.1 | XP_025007765.1 | 1 - 2 | 13952 | 80 | |
XM_025151998.1 | XP_025007766.1 | 2 - 3 | 13415 | 80 | ||
GenBank mRNA | AY753292.1 | - | 2 - 3 | 265 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |