Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293935.2 | XP_015149421.1 | 4 - 5 | 2591 | 55 | |
XM_414607.6 | XP_414607.5 | 4 - 5 | 2590 | 55 | ||
GenBank mRNA | CR389798.1 | - | 2 - 3 | 392 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |