Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422723.6 | XP_422723.3 | 2 - 3 | 524 | 67 | |
GenBank mRNA | CR390246.1 | - | 1 - 2 | 39 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |