Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421829.6 | XP_421829.3 | 5 - 6 | 621 | 77 | |
GenBank mRNA | BX931008.2 | - | 5 - 6 | 615 | 77 | |
BX932000.2 | - | 5 - 6 | 582 | 77 | ||
BX933976.1 | - | 5 - 6 | 560 | 77 | ||
BX933995.1 | - | 5 - 6 | 713 | 77 | ||
BX935850.2 | - | 5 - 6 | 590 | 77 | ||
CR407493.1 | - | 5 - 6 | 618 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |