Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277921.1 | NP_001264850.1 | 4 - 5 | 321 | 130 | |
NM_001277922.1 | NP_001264851.1 | 4 - 5 | 321 | 130 | ||
GenBank mRNA | BU298768.1 | - | 4 - 5 | 311 | 131 | |
CR390466.1 | - | 4 - 5 | 229 | 130 | ||
MF967031.1 | - | 4 - 5 | 335 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |