Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PENK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419213.5 | XP_419213.1 | 2 - 3 | 338 | 71 | |
GenBank mRNA | BX930983.1 | - | 2 - 3 | 317 | 71 | |
CR405789.1 | - | 1 - 2 | 73 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |