Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949515.3 | XP_004949572.1 | 6 - 7 | 622 | 101 | |
XM_004949517.3 | XP_004949574.1 | 5 - 6 | 501 | 101 | ||
XM_015280701.1 | XP_015136187.1 | 6 - 7 | 674 | 101 | ||
XM_015280702.1 | XP_015136188.1 | 7 - 8 | 823 | 101 | ||
XM_015280703.2 | XP_015136189.1 | 6 - 7 | 620 | 101 | ||
XM_015280704.2 | XP_015136190.1 | 6 - 7 | 823 | 101 | ||
XM_025144918.1 | XP_025000686.1 | 5 - 6 | 553 | 101 | ||
XM_424912.6 | XP_424912.3 | 6 - 7 | 584 | 101 | ||
GenBank mRNA | CR524084.1 | - | 4 - 5 | 416 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |