Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198643.1 | NP_001185572.1 | 5 - 6 | 793 | 65 | |
GenBank mRNA | BX929795.1 | - | 5 - 6 | 793 | 65 | |
BX931888.1 | - | 5 - 6 | 574 | 65 | ||
BX950620.2 | - | 2 - 3 | 360 | 65 | ||
CR523233.1 | - | 4 - 5 | 475 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |