Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944787.3 | XP_004944844.2 | 3 - 4 | 258 | 137 | |
XM_025154966.1 | XP_025010734.1 | 3 - 4 | 268 | 137 | ||
XM_025154967.1 | XP_025010735.1 | 3 - 4 | 257 | 137 | ||
XM_414505.6 | XP_414505.3 | 3 - 4 | 268 | 137 | ||
GenBank mRNA | BX930082.1 | - | 3 - 4 | 257 | 137 | |
BX936052.1 | - | 3 - 4 | 269 | 137 | ||
BX936106.1 | - | 3 - 4 | 269 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |