Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417606.5 | XP_417606.4 | 3 - 4 | 407 | 134 | |
GenBank mRNA | BX929289.2 | - | 3 - 4 | 372 | 134 | |
BX931240.1 | - | 3 - 4 | 377 | 134 | ||
BX934511.1 | - | 3 - 4 | 422 | 134 | ||
BX934521.1 | - | 3 - 4 | 390 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |