Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277399.1 | NP_001264328.1 | 4 - 5 | 395 | 101 | |
GenBank mRNA | BX934448.1 | - | 4 - 5 | 361 | 101 | |
CR353120.1 | - | 4 - 5 | 368 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |