Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935278.2 | XP_004935335.1 | 3 - 4 | 239 | 92 | |
XM_419377.6 | XP_419377.1 | 3 - 4 | 238 | 92 | ||
GenBank mRNA | BX932118.1 | - | 3 - 4 | 187 | 92 | |
BX934462.2 | - | 3 - 4 | 186 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |