Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277886.1 | NP_001264815.1 | 4 - 5 | 373 | 98 | |
XM_015278223.2 | XP_015133709.1 | 4 - 5 | 388 | 98 | ||
XM_015278224.2 | XP_015133710.1 | 4 - 5 | 389 | 98 | ||
XM_025149547.1 | XP_025005315.1 | 4 - 5 | 389 | 98 | ||
GenBank mRNA | BU112689.1 | - | 4 - 5 | 374 | 98 | |
BX930950.1 | - | 4 - 5 | 359 | 98 | ||
BX935900.2 | - | 4 - 5 | 375 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |