Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949515.3 | XP_004949572.1 | 15 - 16 | 1660 | 72 | |
XM_004949517.3 | XP_004949574.1 | 14 - 15 | 1539 | 72 | ||
XM_015280701.1 | XP_015136187.1 | 15 - 16 | 1712 | 72 | ||
XM_015280702.1 | XP_015136188.1 | 16 - 17 | 1861 | 72 | ||
XM_015280703.2 | XP_015136189.1 | 15 - 16 | 1658 | 72 | ||
XM_015280704.2 | XP_015136190.1 | 15 - 16 | 1861 | 72 | ||
XM_025144918.1 | XP_025000686.1 | 14 - 15 | 1591 | 72 | ||
XM_424912.6 | XP_424912.3 | 15 - 16 | 1622 | 72 | ||
GenBank mRNA | CR385816.1 | - | 8 - 9 | 810 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |