Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298756.2 | XP_015154242.1 | 1050 | 75 | ||
XM_015298757.2 | XP_015154243.1 | 985 | 75 | |||
XM_015298758.2 | XP_015154244.1 | 986 | 75 | |||
XM_015298759.2 | XP_015154245.1 | 562 | 75 | |||
GenBank mRNA | U46127.1 | - | 1017 | 75 | ||
U46128.2 | - | 1013 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus |