Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277492.1 | NP_001264421.1 | 4 - 5 | 1046 | 141 | |
GenBank mRNA | BX935008.1 | - | 4 - 5 | 1040 | 141 | |
KU180444.1 | - | 2 - 3 | 917 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |