Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278089.1 | NP_001265018.1 | 4 - 5 | 682 | 82 | |
GenBank mRNA | BX950347.1 | - | 4 - 5 | 672 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |