Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276338.1 | NP_001263267.1 | 6 - 7 | 511 | 85 | |
GenBank mRNA | BU204597.1 | - | 5 - 6 | 280 | 85 | |
BU225566.1 | - | 6 - 7 | 511 | 85 | ||
BU324297.1 | - | 5 - 6 | 454 | 85 | ||
CR352660.1 | - | 5 - 6 | 300 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |