Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199585.1 | NP_001186514.1 | 1 - 2 | 49 | 89 | |
XM_015281377.2 | XP_015136863.1 | 1 - 2 | 225 | 89 | ||
XM_015281378.2 | XP_015136864.1 | 1 - 2 | 225 | 89 | ||
GenBank mRNA | BX932120.2 | - | 1 - 2 | 49 | 89 | |
CR405818.1 | - | 1 - 2 | 65 | 89 | ||
CR405822.1 | - | 1 - 2 | 66 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |