Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199585.1 | NP_001186514.1 | 2 - 3 | 156 | 136 | |
XM_015281377.2 | XP_015136863.1 | 2 - 3 | 332 | 136 | ||
XM_015281378.2 | XP_015136864.1 | 2 - 3 | 332 | 136 | ||
GenBank mRNA | BX931729.1 | - | 2 - 3 | 149 | 136 | |
BX932120.2 | - | 2 - 3 | 156 | 136 | ||
CR405818.1 | - | 2 - 3 | 172 | 136 | ||
CR405822.1 | - | 2 - 3 | 173 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |