Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277824.1 | NP_001264753.1 | 1 - 2 | 65 | 72 | |
GenBank mRNA | AM067556.1 | - | 1 - 2 | 65 | 72 | |
BX934467.2 | - | 1 - 2 | 38 | 72 | ||
BX936078.1 | - | 1 - 2 | 57 | 72 | ||
BX936226.2 | - | 1 - 2 | 66 | 72 | ||
CN219349.1 | - | 1 - 2 | 53 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |