Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277535.1 | NP_001264464.1 | 3 - 4 | 526 | 67 | |
GenBank mRNA | BX931304.1 | - | 3 - 4 | 508 | 67 | |
BX931763.2 | - | 1 - 2 | 512 | 67 | ||
BX933243.1 | - | 1 - 2 | 529 | 67 | ||
BX933732.2 | - | 1 - 2 | 588 | 67 | ||
BX934499.2 | - | 3 - 4 | 526 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |