Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199584.1 | NP_001186513.1 | 8 - 9 | 1491 | 126 | |
XM_015286372.2 | XP_015141858.1 | 9 - 10 | 1675 | 126 | ||
XM_015286373.2 | XP_015141859.1 | 12 - 13 | 3481 | 126 | ||
XM_025150668.1 | XP_025006436.1 | 9 - 10 | 1680 | 126 | ||
XM_025150669.1 | XP_025006437.1 | 12 - 13 | 3481 | 126 | ||
GenBank mRNA | BX934411.1 | - | 5 - 6 | 698 | 126 | |
BX936092.2 | - | 4 - 5 | 538 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |