Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004948703.3 | XP_004948760.1 | 6 - 7 | 1022 | 68 | |
XM_418193.6 | XP_418193.3 | 6 - 7 | 1003 | 68 | ||
GenBank mRNA | CR352713.1 | - | 4 - 5 | 448 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |