Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293321.1 | NP_001280250.1 | 2 - 3 | 573 | 77 | |
XM_025154160.1 | XP_025009928.1 | 1 - 2 | 159 | 77 | ||
XM_025154161.1 | XP_025009929.1 | 2 - 3 | 272 | 77 | ||
GenBank mRNA | CR353629.1 | - | 2 - 3 | 588 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |