Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284545.2 | XP_015140031.1 | 4 | 1791 | 117 | |
XM_015284546.2 | XP_015140032.1 | 4 | 1667 | 117 | ||
XM_015284547.2 | XP_015140033.1 | 3 | 1566 | 117 | ||
GenBank mRNA | BX934207.2 | - | 1 | 1050 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |