Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277884.1 | NP_001264813.1 | 2 - 3 | 201 | 154 | |
GenBank mRNA | BX929622.1 | - | 2 - 3 | 201 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |