Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX930771.1 | - | 3 - 4 | 274 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |