Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PENK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419213.5 | XP_419213.1 | 1 - 2 | 197 | 133 | |
GenBank mRNA | BX930983.1 | - | 1 - 2 | 176 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |