Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288649.2 | XP_015144135.1 | 2 - 3 | 204 | 67 | |
GenBank mRNA | BX931374.1 | - | 2 - 3 | 187 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |