Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414535.6 | XP_414535.3 | 4 - 5 | 892 | 81 | |
GenBank mRNA | CR523593.1 | - | 3 - 4 | 478 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |