Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005798.1 | NP_001005798.1 | 3 - 4 | 1003 | 77 | |
XM_015283067.2 | XP_015138553.2 | 3 - 4 | 1072 | 77 | ||
GenBank mRNA | AJ719936.1 | - | 3 - 4 | 1003 | 77 | |
CR388496.1 | - | 3 - 4 | 623 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |