Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006345.2 | NP_001006345.1 | 6 - 7 | 773 | 97 | |
GenBank mRNA | AJ720578.1 | - | 6 - 7 | 767 | 97 | |
CR338953.1 | - | 6 - 7 | 753 | 97 | ||
CR352384.1 | - | 5 - 6 | 743 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |