Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006471.1 | NP_001006471.1 | 1 - 2 | 153 | 83 | |
XM_015286612.2 | XP_015142098.1 | 1 - 2 | 157 | 83 | ||
GenBank mRNA | AJ720279.1 | - | 1 - 2 | 153 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |