Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204549.1 | NP_989880.1 | 6 - 7 | 641 | 99 | |
XM_015291368.2 | XP_015146854.1 | 6 - 7 | 663 | 99 | ||
GenBank mRNA | AF515726.1 | - | 6 - 7 | 641 | 99 | |
AJ720280.1 | - | 6 - 7 | 663 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |