Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204558.1 | NP_989889.1 | 4 - 5 | 548 | 84 | |
XM_025142159.1 | XP_024997927.1 | 4 - 5 | 849 | 84 | ||
GenBank mRNA | AF380350.1 | - | 4 - 5 | 548 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |