Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204818.2 | NP_990149.1 | 4 - 5 | 419 | 68 | |
XM_015284816.2 | XP_015140302.1 | 5 - 6 | 518 | 68 | ||
GenBank mRNA | AF133251.1 | - | 4 - 5 | 422 | 68 | |
KP686393.1 | - | 4 - 5 | 409 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |