Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204819.1 | NP_990150.1 | 4 - 5 | 507 | 100 | |
GenBank mRNA | J05643.2 | - | 4 - 5 | 507 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |