Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204834.1 | NP_990165.1 | 1 - 2 | 923 | 111 | |
XM_015280910.2 | XP_015136396.1 | 2 - 3 | 326 | 111 | ||
GenBank mRNA | AF160960.1 | - | 1 - 2 | 923 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |