Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204869.1 | NP_990200.1 | 2 - 3 | 342 | 80 | |
XM_015278760.2 | XP_015134246.1 | 2 - 3 | 338 | 80 | ||
GenBank mRNA | AF129933.1 | - | 2 - 3 | 342 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |