Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204869.1 | NP_990200.1 | 7 - 8 | 819 | 68 | |
GenBank mRNA | AF129933.1 | - | 7 - 8 | 819 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |