Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204878.1 | NP_990209.1 | 8 - 9 | 1028 | 79 | |
XM_025150584.1 | XP_025006352.1 | 8 - 9 | 1075 | 79 | ||
XM_025150585.1 | XP_025006353.1 | 8 - 9 | 1076 | 79 | ||
GenBank mRNA | AB025281.1 | - | 8 - 9 | 1028 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |