Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204881.1 | NP_990212.1 | 3 - 4 | 912 | 65 | |
GenBank mRNA | AJ012219.1 | - | 3 - 4 | 912 | 65 | |
U41445.2 | - | 3 - 4 | 342 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |