Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204889.1 | NP_990220.1 | 4 - 5 | 916 | 83 | |
XM_015279326.2 | XP_015134812.2 | 4 - 5 | 1863 | 83 | ||
GenBank mRNA | AB005882.1 | - | 4 - 5 | 916 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |