Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205469.1 | NP_990800.1 | 3 - 4 | 319 | 68 | |
XM_015278795.2 | XP_015134281.1 | 3 - 4 | 321 | 68 | ||
XM_015278796.2 | XP_015134282.1 | 4 - 5 | 381 | 68 | ||
XM_025153808.1 | XP_025009576.1 | 4 - 5 | 381 | 68 | ||
GenBank mRNA | J00903.1 | - | 3 - 4 | 319 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |