Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205469.1 | NP_990800.1 | 9 - 10 | 1378 | 111 | |
XM_015278796.2 | XP_015134282.1 | 10 - 11 | 1440 | 111 | ||
GenBank mRNA | J00903.1 | - | 9 - 10 | 1378 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |