Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005816.1 | NP_001005816.1 | 5 - 6 | 639 | 99 | |
NM_001012622.1 | NP_001012640.1 | 5 - 6 | 647 | 99 | ||
GenBank mRNA | AB188512.1 | - | 5 - 6 | 647 | 99 | |
AB188513.1 | - | 4 - 5 | 548 | 99 | ||
AJ719707.1 | - | 5 - 6 | 639 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |